Methyl [1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate

Supplier: Matrix Scientific

080792-500MG 080792-1G
102671-248EA 258.46 USD
102671-248 102671-250
Methyl [1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate
Methyl [1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD03409104
CAS Number: 524051-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR