N-[4-(Acetylamino)phenyl]-2-[1-(chloroacetyl)-3-oxopiperazin-2-yl]acetamide

Supplier: Matrix Scientific

080791-500MG 080791-1G
102671-244EA 258.46 USD
102671-244 102671-246
N-[4-(Acetylamino)phenyl]-2-[1-(chloroacetyl)-3-oxopiperazin-2-yl]acetamide
N-[4-(Acetylamino)phenyl]-2-[1-(chloroacetyl)-3-oxopiperazin-2-yl]acetamide

Hazard: Irritant

Formula: C₁₆H₁₉ClN₄O₄
MDL Number: MFCD22374960
CAS Number: 1379811-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR