Tetraethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Supplier: Matrix Scientific

080785-500MG 080785-1G
102671-230EA 161.54 USD
102671-230 102671-232
Tetraethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Tetraethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Hazard: Irritant

Formula: C₂₉H₃₃NO₈S₃
MW: 619.78 g/mol
MDL Number: MFCD00521284
CAS Number: 296272-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR