Tetramethyl-5',5',8',9'-tetramethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Supplier: Matrix Scientific

080780-500MG 080780-1G 080780-5G
102671-208EA 161.54 USD
102671-208 102671-210 102671-212
Tetramethyl-5',5',8',9'-tetramethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Tetramethyl-5',5',8',9'-tetramethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Hazard: Irritant

Formula: C₂₆H₂₇NO₈S₃
MW: 577.7 g/mol
MDL Number: MFCD01036904
CAS Number: 331761-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR