Tetramethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Supplier: Matrix Scientific

080779-500MG 080779-1G
102671-204EA 161.54 USD
102671-204 102671-206
Tetramethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Tetramethyl-5',5',9'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate

Hazard: Irritant

Formula: C₂₅H₂₅NO₈S₃
MW: 563.67 g/mol
MDL Number: MFCD00475728
CAS Number: 258267-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR