Diethyl-2-(2,2,6-trimethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate

Supplier: Matrix Scientific

080776-500MG 080776-1G
102671-190EA 193.85 USD
102671-190 102671-192
Diethyl-2-(2,2,6-trimethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate
Diethyl-2-(2,2,6-trimethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₂₁H₂₃NO₄S₃
MDL Number: MFCD00532507
CAS Number: 296272-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR