Dimethyl-2-(2,2,6,7-tetramethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate

Supplier: Matrix Scientific

080772-500MG 080772-1G 080772-5G
102671-170EA 193.85 USD
102671-170 102671-172 102671-174
Dimethyl-2-(2,2,6,7-tetramethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate
Dimethyl-2-(2,2,6,7-tetramethyl-3-thioxo-2,3-dihydroquinolin-4(1H)-ylidene)-1,3-dithiole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₂₀H₂₁NO₄S₃
MDL Number: MFCD01138051
CAS Number: 330179-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR