2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol

Supplier: Matrix Scientific

Synonyms: 2-(2-Benzoxazolylamino)-6-methylpyrimidin-4(3H)-one

080769-500MG 080769-1G
102671-160EA 258.46 USD
102671-160 102671-162
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol

Hazard: Irritant

Formula: C₁₂H₁₀N₄O₂
MW: 242.24 g/mol
MDL Number: MFCD01948713
CAS Number: 86328-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR