2-Methyl-3,5,7-triphenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine

Supplier: Matrix Scientific

080768-500MG 080768-1G
102671-156EA 193.85 USD
102671-156 102671-158
2-Methyl-3,5,7-triphenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine
2-Methyl-3,5,7-triphenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₂₅H₂₁N₃
MDL Number: MFCD22374954
CAS Number: 1379811-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR