1-(1,3-Benzothiazol-2-yl)pentane-2,4-dione

Supplier: Matrix Scientific

080765-500MG 080765-1G
102671-144EA 387.69 USD
102671-144 102671-146
1-(1,3-Benzothiazol-2-yl)pentane-2,4-dione
1-(1,3-Benzothiazol-2-yl)pentane-2,4-dione

Hazard: Irritant

Formula: C₁₂H₁₁NO₂S
MDL Number: MFCD22374953
CAS Number: 1379811-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR