1,1'-(6-Chloro-1,3,5-triazine-2,4-diyl)-bis-1,2,3,4-tetrahydroquinoline
Supplier: Matrix Scientific
080762-500MG
080762-1G
102671-134EA
193.85
USD
102671-134
102671-136
1,1'-(6-Chloro-1,3,5-triazine-2,4-diyl)-bis-1,2,3,4-tetrahydroquinoline
1,1'-(6-Chloro-1,3,5-triazine-2,4-diyl)-bis-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₂₁H₂₀ClN₅ |
MDL Number:
MFCD03296803 CAS Number: 489422-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...