6-Chloro-N,N'-bis(4-ethoxyphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080758-500MG 080758-1G 080758-5G
102671-114EA 193.85 USD
102671-114 102671-116 102671-118
6-Chloro-N,N'-bis(4-ethoxyphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(4-ethoxyphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₉H₂₀ClN₅O₂
MDL Number: MFCD01951678
CAS Number: 78912-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR