6-Chloro-N,N'-bis(2,4-dimethoxyphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080756-500MG 080756-1G 080756-5G
102671-102EA 193.85 USD
102671-102 102671-104 102671-106
6-Chloro-N,N'-bis(2,4-dimethoxyphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(2,4-dimethoxyphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₉H₂₀ClN₅O₄
MDL Number: MFCD02733211
CAS Number: 333326-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR