6-Chloro-N,N'-bis(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine
Supplier: Matrix Scientific
080754-500MG
080754-1G
102671-094EA
193.85
USD
102671-094
102671-096
6-Chloro-N,N'-bis(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine
Hazard: Irritant
Formula:
C₁₇H₁₆ClN₅O₂ |
MDL Number:
MFCD03225379 CAS Number: 62752-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...