6-Chloro-N,N'-bis(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080750-500MG 080750-1G
102671-076EA 193.85 USD
102671-076 102671-078
6-Chloro-N,N'-bis(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₉H₂₀ClN₅
MDL Number: MFCD01952265
CAS Number: 374101-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR