6-Chloro-N,N'-di-1-naphthyl-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080748-500MG 080748-1G 080748-5G
102671-064EA 193.85 USD
102671-064 102671-066 102671-068
6-Chloro-N,N'-di-1-naphthyl-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-di-1-naphthyl-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₂₃H₁₆ClN₅
MDL Number: MFCD22374952
CAS Number: 30355-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR