6-Chloro-N,N'-bis(2-ethylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080745-500MG 080745-1G
102671-052EA 193.85 USD
102671-052 102671-054
6-Chloro-N,N'-bis(2-ethylphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(2-ethylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₉H₂₀ClN₅
MDL Number: MFCD02239496
CAS Number: 364748-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR