6-Chloro-N,N'-bis(3,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080742-500MG 080742-1G 080742-5G
102671-036EA 193.85 USD
102671-036 102671-038 102671-040
6-Chloro-N,N'-bis(3,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(3,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₅H₈Cl₅N₅
MDL Number: MFCD01953481
CAS Number: 30355-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR