6-Chloro-N,N'-bis(4-isopropylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080740-500MG 080740-1G
102671-028EA 193.85 USD
102671-028 102671-030
6-Chloro-N,N'-bis(4-isopropylphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(4-isopropylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₂₁H₂₄ClN₅
MDL Number: MFCD02817174
CAS Number: 377067-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR