6-(Chloromethyl)-N-(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080733-500MG 080733-1G
102671-002EA 258.46 USD
102671-002 102671-004
6-(Chloromethyl)-N-(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(2,4-dimethylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₅
MDL Number: MFCD03190397
CAS Number: 444081-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR