6-(Chloromethyl)-N-(2-methoxy-5-methylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080726-500MG 080726-1G
102670-974EA 258.46 USD
102670-974 102670-976
6-(Chloromethyl)-N-(2-methoxy-5-methylphenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(2-methoxy-5-methylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₅O
MW: 279.73 g/mol
MDL Number: MFCD22374948
CAS Number: 1379811-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR