6-(Chloromethyl)-N-(2,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine
Supplier: Matrix Scientific
080722-500MG
080722-1G
080722-5G
102670-954EA
258.46
USD
102670-954
102670-956
102670-958
6-(Chloromethyl)-N-(2,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(2,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine
Hazard: Irritant
Formula:
C₁₀H₈Cl₃N₅ |
MDL Number:
MFCD22374945 CAS Number: 1379811-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...