N-(4-Phenoxyphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080718-500MG 080718-1G 080718-5G
102670-940EA 193.85 USD
102670-940 102670-942 102670-944
N-(4-Phenoxyphenyl)imidodicarbonimidic diamide
N-(4-Phenoxyphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₁₄H₁₅N₅O
MDL Number: MFCD03321766
CAS Number: 92295-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR