N-Biphenyl-2-ylimidodicarbonimidic diamide
Supplier: Matrix Scientific
080717-500MG
080717-1G
102670-936EA
193.85
USD
102670-936
102670-938
N-Biphenyl-2-ylimidodicarbonimidic diamide
N-Biphenyl-2-ylimidodicarbonimidic diamide
Hazard: Irritant
Formula:
C₁₄H₁₅N₅ |
MDL Number:
MFCD07783240 CAS Number: 10378-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...