Dimethyl-5-{[{[amino(imino)methyl]amino}(imino)methyl]amino}isophthalate

Supplier: Matrix Scientific

080716-500MG 080716-1G
102670-932EA 193.85 USD
102670-932 102670-934
Dimethyl-5-{[{[amino(imino)methyl]amino}(imino)methyl]amino}isophthalate
Dimethyl-5-{[{[amino(imino)methyl]amino}(imino)methyl]amino}isophthalate

Hazard: Irritant

Formula: C₁₂H₁₅N₅O₄
MDL Number: MFCD03615413
CAS Number: 524055-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR