N-(3-Nitrophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080715-500MG 080715-1G
102670-928EA 193.85 USD
102670-928 102670-930
N-(3-Nitrophenyl)imidodicarbonimidic diamide
N-(3-Nitrophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₈H₁₀N₆O₂
MDL Number: MFCD00448708
CAS Number: 10441-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR