N-(4-Methyl-3-nitrophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080714-500MG 080714-1G
102670-924EA 193.85 USD
102670-924 102670-926
N-(4-Methyl-3-nitrophenyl)imidodicarbonimidic diamide
N-(4-Methyl-3-nitrophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₉H₁₂N₆O₂
MDL Number: MFCD22374943
CAS Number: 1379811-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR