N-(4-Bromophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080713-500MG 080713-1G
102670-920EA 193.85 USD
102670-920 102670-922
N-(4-Bromophenyl)imidodicarbonimidic diamide
N-(4-Bromophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₈H₁₀BrN₅
MDL Number: MFCD00464642
CAS Number: 101250-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR