N-(2,3-Dichlorophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080712-500MG 080712-1G
102670-916EA 193.85 USD
102670-916 102670-918
N-(2,3-Dichlorophenyl)imidodicarbonimidic diamide
N-(2,3-Dichlorophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₈H₉Cl₂N₅
MDL Number: MFCD00276882
CAS Number: 143413-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR