N-(3,5-Dichlorophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080711-500MG 080711-1G 080711-5G
102670-910EA 193.85 USD
102670-910 102670-912 102670-914
N-(3,5-Dichlorophenyl)imidodicarbonimidic diamide
N-(3,5-Dichlorophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₈H₉Cl₂N₅
MDL Number: MFCD00179295
CAS Number: 1672-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR