N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific
Warning

Synonyms: 1-Carbamimidamido-N-(2,4-dichlorophenyl)methanimidamide

080710-500MG 080710-1G 080710-5G
102670-904EA 193.85 USD
102670-904 102670-906 102670-908
N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide
N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₈H₁₀Cl₃N₅
MW: 246.10 g/mol
MDL Number: MFCD00448709
CAS Number: 101252-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR