N-(5-Chloro-2-methylphenyl)imidodicarbonimidicdiamide

Supplier: Matrix Scientific

080709-500MG 080709-1G 080709-5G
102670-898EA 258.46 USD
102670-898 102670-900 102670-902
N-(5-Chloro-2-methylphenyl)imidodicarbonimidicdiamide
N-(5-Chloro-2-methylphenyl)imidodicarbonimidicdiamide

Hazard: Irritant

Formula: C₉H₁₂ClN₅
MW: 225.68 g/mol
MDL Number: MFCD03615418
CAS Number: 49872-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR