N-(3-Methylphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080707-500MG 080707-1G 080707-5G
102670-886EA 193.85 USD
102670-886 102670-888 102670-890
N-(3-Methylphenyl)imidodicarbonimidic diamide
N-(3-Methylphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₉H₁₃N₅
MDL Number: MFCD00456189
CAS Number: 46338-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR