N-(2-Methoxy-5-methylphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080705-500MG 080705-1G 080705-5G
102670-874EA 193.85 USD
102670-874 102670-876 102670-878
N-(2-Methoxy-5-methylphenyl)imidodicarbonimidic diamide
N-(2-Methoxy-5-methylphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₁₀H₁₅N₅O
MW: 221.26 g/mol
MDL Number: MFCD22374942
CAS Number: 1379811-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR