N-(4-Methoxyphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080704-500MG 080704-1G
102670-870EA 193.85 USD
102670-870 102670-872
N-(4-Methoxyphenyl)imidodicarbonimidic diamide
N-(4-Methoxyphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₉H₁₃N₅O
MDL Number: MFCD00448707
CAS Number: 43191-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR