N-(2-Ethoxyphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080703-500MG 080703-1G
102670-866EA 193.85 USD
102670-866 102670-868
N-(2-Ethoxyphenyl)imidodicarbonimidic diamide
N-(2-Ethoxyphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₁₀H₁₅N₅O
MDL Number: MFCD03615408
CAS Number: 524055-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR