5-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Supplier: Matrix Scientific

080699-500MG 080699-1G 080699-5G
102670-844EA 775.38 USD
102670-844 102670-846 102670-848
5-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride
5-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₀Cl₂N₂
MW: 181.07 g/mol
MDL Number: MFCD22421992
CAS Number: 1417568-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR