5-Methyl-1-propyl-1H-pyrazole-3-carboxylic acid
Supplier: Matrix Scientific
080696-500MG
080696-1G
080696-5G
102670-826EA
516.92
USD
102670-826
102670-828
102670-830
5-Methyl-1-propyl-1H-pyrazole-3-carboxylic acid
5-Methyl-1-propyl-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₂N₂O₂ |
MDL Number:
MFCD08700640 CAS Number: 956950-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...