Ethyl-4-iodo-3,5-dimethyl-1H-pyrazole-1-carboxylate
Supplier: Matrix Scientific
080695-500MG
080695-1G
080695-5G
102670-820EA
387.69
USD
102670-820
102670-822
102670-824
Ethyl-4-iodo-3,5-dimethyl-1H-pyrazole-1-carboxylate
Ethyl-4-iodo-3,5-dimethyl-1H-pyrazole-1-carboxylate
Hazard: Irritant
Formula:
C₈H₁₁IN₂O₂ |
MDL Number:
MFCD22374941 CAS Number: 1379811-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...