1,2-Dimethyl-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

080676-500MG 080676-1G 080676-5G
102670-724EA 261.54 USD
102670-724 102670-726 102670-728
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₉NO₂
MDL Number: MFCD19230065
CAS Number: 89776-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR