8-(Benzyloxy)-2-methylquinoline

Supplier: Matrix Scientific

080667-500MG 080667-1G 080667-5G
102670-670EA 193.85 USD
102670-670 102670-672 102670-674
8-(Benzyloxy)-2-methylquinoline
8-(Benzyloxy)-2-methylquinoline

Hazard: Irritant

Formula: C₁₇H₁₅NO
MDL Number: MFCD00169012
CAS Number: 93315-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR