Diethyl (8-hydroxyquinolin-2-yl)malonate
Supplier: Matrix Scientific
080663-500MG
080663-1G
080663-5G
102670-646EA
193.85
USD
102670-646
102670-648
102670-650
Diethyl (8-hydroxyquinolin-2-yl)malonate
Diethyl (8-hydroxyquinolin-2-yl)malonate
Hazard: Irritant
Formula:
C₁₆H₁₇NO₅ |
MDL Number:
MFCD00168971 CAS Number: 36018-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...