[2-(Cyanoamino)-4-hydroxypyrimidin-5-yl]acetic acid hydrochloride

Supplier: Matrix Scientific

080652-500MG 080652-1G
102670-588EA 387.69 USD
102670-588 102670-590
[2-(Cyanoamino)-4-hydroxypyrimidin-5-yl]acetic acid hydrochloride
[2-(Cyanoamino)-4-hydroxypyrimidin-5-yl]acetic acid hydrochloride

Hazard: Irritant

Formula: C₇H₇ClN₄O₃
MDL Number: MFCD22421991
CAS Number: 1417567-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR