4-(4-Isopropylphenyl)-2,5-dimethyl-3-furoic acid

Supplier: Matrix Scientific

080647-500MG 080647-1G
102670-562EA 516.92 USD
102670-562 102670-564
4-(4-Isopropylphenyl)-2,5-dimethyl-3-furoic acid
4-(4-Isopropylphenyl)-2,5-dimethyl-3-furoic acid

Hazard: Irritant

Formula: C₁₆H₁₈O₃
MDL Number: MFCD22374936
CAS Number: 1379811-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR