2,5-Dimethyl-4-phenyl-3-furoic acid

Supplier: Matrix Scientific

080645-500MG 080645-1G 080645-5G
102670-552EA 387.69 USD
102670-552 102670-554 102670-556
2,5-Dimethyl-4-phenyl-3-furoic acid
2,5-Dimethyl-4-phenyl-3-furoic acid

Hazard: Irritant

Formula: C₁₃H₁₂O₃
MDL Number: MFCD00684731
CAS Number: 104794-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR