Ethyl-3-amino-5-(3-chlorophenyl)thiophene-2-carboxylate

Supplier: Matrix Scientific

080641-500MG 080641-1G 080641-5G
102670-528EA 258.46 USD
102670-528 102670-530 102670-532
Ethyl-3-amino-5-(3-chlorophenyl)thiophene-2-carboxylate
Ethyl-3-amino-5-(3-chlorophenyl)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₂S
MDL Number: MFCD09835655
CAS Number: 1094398-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR