Ethyl-3-amino-5-(4-cyclohexylphenyl)thiophene-2-carboxylate

Supplier: Matrix Scientific

080640-500MG 080640-1G 080640-5G
102670-522EA 258.46 USD
102670-522 102670-524 102670-526
Ethyl-3-amino-5-(4-cyclohexylphenyl)thiophene-2-carboxylate
Ethyl-3-amino-5-(4-cyclohexylphenyl)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₃NO₂S
MDL Number: MFCD22374935
CAS Number: 1379811-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR