Ethyl-3-amino-5-(4-ethylphenyl)thiophene-2-carboxylate

Supplier: Matrix Scientific

080639-500MG 080639-1G 080639-5G
102670-516EA 258.46 USD
102670-516 102670-518 102670-520
Ethyl-3-amino-5-(4-ethylphenyl)thiophene-2-carboxylate
Ethyl-3-amino-5-(4-ethylphenyl)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₂S
MDL Number: MFCD12540730
CAS Number: 1096972-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR