Ethyl-3-amino-5-(3-methylphenyl)thiophene-2-carboxylate
Supplier: Matrix Scientific
080638-500MG
080638-1G
080638-5G
102670-510EA
258.46
USD
102670-510
102670-512
102670-514
Ethyl-3-amino-5-(3-methylphenyl)thiophene-2-carboxylate
Ethyl-3-amino-5-(3-methylphenyl)thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₅NO₂S MW: 261.35 g/mol |
MDL Number:
MFCD09835651 CAS Number: 1019012-08-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...