(2E)-4-(Biphenyl-2-ylamino)-4-oxobut-2-enoic acid

Supplier: Matrix Scientific

080636-500MG 080636-1G
102670-502EA 193.85 USD
102670-502 102670-504
(2E)-4-(Biphenyl-2-ylamino)-4-oxobut-2-enoic acid
(2E)-4-(Biphenyl-2-ylamino)-4-oxobut-2-enoic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD01137706
CAS Number: 1164564-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR